Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr. 1: 39955112 - 39969968 on Build GRCh38
UniGene ID
Hs.655177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136493.2 NP_001129965.1 12 1461
NM_001287808.1 NP_001274737.1 10 1162
NM_001287809.1 NP_001274738.1 12 1305
NM_032793.4 NP_116182.2 12 1422
NR_109896.1 10 1225
XM_005271285.1 XP_005271342.1 12 1386
XM_011542312.2 XP_011540614.1 11 1305
GenBank mRNA AB073383.1 - 12 1386
AF289609.1 - 12 1378
AF370364.1 - 7 1682
AK074186.1 - 12 1327
AK075183.1 - 12 1389
AK093223.1 - 12 1423
AK291540.1 - 12 1384
AK297991.1 - 12 1269
AK301533.1 - 10 1129
AL834184.1 - 10 1179
AY358636.1 - 12 1404
BC006353.2 - 8 757
BC011587.1 - 12 1396 4646430
BC092414.1 - 12 1364 30387073

View Full Product Details