Gene Transcripts

Gene Symbol
CALML6
Entrez Gene ID
163688
Gene Name
calmodulin like 6
Gene Aliases
CAGLP
Chromosome Location
Chr. 1: 1913751 - 1917294 on Build GRCh38
UniGene ID
Hs.85902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138705.2 NP_619650.2 1, 2 474
GenBank mRNA AF490905.2 - 1, 2 474
AY566229.1 - 1, 2 474

View Full Product Details