Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178819.3 | NP_848934.1 | 4 | 1337 | |
XM_011544392.2 | XP_011542694.1 | 5 | 1460 | ||
XM_011544393.1 | XP_011542695.1 | 4 | 1349 | ||
XM_011544394.1 | XP_011542696.1 | 4 | 1386 | ||
XM_017013042.1 | XP_016868531.1 | 5 | 1487 | ||
XM_017013043.1 | XP_016868532.1 | 4 | 1335 | ||
GenBank mRNA | AF317516.1 | - | 3 | 563 | |
AF406612.1 | - | 4 | 1325 | ||
AY358670.1 | - | 3 | 474 | ||
AY513610.1 | - | 3 | 557 | ||
BC051377.1 | - | 3 | 517 | 6649895 | |
BC061884.1 | - | 3 | 517 | 6577569 |