Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282590.1 | NP_001269519.1 | 2 | 406 | |
NM_001282591.1 | NP_001269520.1 | 2 | 406 | ||
NM_001282592.1 | NP_001269521.1 | 3 | 727 | ||
NM_001282593.1 | NP_001269522.1 | 2 | 406 | ||
NM_001282594.1 | NP_001269523.1 | 2 | 406 | ||
NM_001282595.1 | NP_001269524.1 | 2 | 188 | ||
NM_001282596.1 | NP_001269525.1 | 2 | 406 | ||
NM_021181.4 | NP_067004.3 | 3 | 727 | ||
XM_011509828.1 | XP_011508130.1 | 3 | 537 | ||
XM_011509829.1 | XP_011508131.1 | 3 | 537 | ||
GenBank mRNA | AB027233.1 | - | 3 | 497 | |
AF291815.1 | - | 3 | 493 | ||
AF390894.1 | - | 3 | 502 | ||
AJ271869.1 | - | 3 | 515 | ||
AJ276429.2 | - | 3 | 515 | ||
AK290706.1 | - | 3 | 511 | ||
AK292097.1 | - | 2 | 190 | ||
AK292148.1 | - | 3 | 511 | ||
AK301137.1 | - | 2 | 197 | ||
AK301432.1 | - | 2 | 192 | ||
AK301438.1 | - | 2 | 194 | ||
AL834424.1 | - | 2 | 195 | ||
AY358512.1 | - | 3 | 519 | ||
BC027867.1 | - | 3 | 493 | 5203638 | |
DA978435.1 | - | 2 | 189 | ||
DA978525.1 | - | 2 | 169 |