Gene Transcripts
- Gene Symbol
- CLSPN
- Entrez Gene ID
- 63967
- Gene Name
- claspin
- Gene Aliases
- Chromosome Location
- Chr. 1: 35720465 - 35769985 on Build GRCh38
- UniGene ID
- Hs.175613
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190481.1 |
NP_001177410.1
|
2, 3 |
218 |
|
NM_022111.3 |
NP_071394.2
|
2, 3 |
218 |
|
XM_011541938.2 |
XP_011540240.1
|
2, 3 |
241 |
|
XM_011541939.2 |
XP_011540241.1
|
2, 3 |
235 |
|
XM_011541940.2 |
XP_011540242.1
|
2, 3 |
235 |
|
XM_011541941.2 |
XP_011540243.1
|
2, 3 |
235 |
|
GenBank mRNA |
AF297866.1 |
- |
2, 3 |
218 |
|
AK308193.1 |
- |
2, 3 |
246 |
|
BC018670.1 |
- |
2, 3 |
238 |
|
BC038991.2 |
- |
2, 3 |
244 |
|
BC062215.1 |
- |
2, 3 |
241 |
|
BC113116.1 |
- |
2, 3 |
211 |
7961377 |
BC115025.1 |
- |
2, 3 |
211 |
7961363 |
BC115026.1 |
- |
2, 3 |
211 |
7961366 |
BC137279.1 |
- |
2, 3 |
197 |
9021276 |
BC140789.1 |
- |
2, 3 |
197 |
9021657 |
View Full Product Details