Gene Transcripts
- Gene Symbol
- SIL1
- Entrez Gene ID
- 64374
- Gene Name
- SIL1 nucleotide exchange factor
- Gene Aliases
- BAP, MSS, ULG5
- Chromosome Location
- Chr. 5: 138946720 - 139198376 on Build GRCh38
- UniGene ID
- Hs.483521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037633.1 |
NP_001032722.1
|
3, 4 |
322 |
|
NM_022464.4 |
NP_071909.1
|
2, 3 |
234 |
|
XM_011543570.2 |
XP_011541872.1
|
4, 5 |
359 |
|
GenBank mRNA |
AF547994.1 |
- |
2, 3 |
153 |
|
AJ299442.1 |
- |
2, 3 |
212 |
|
AK074624.1 |
- |
2, 3 |
226 |
|
AK075177.1 |
- |
2, 3 |
163 |
|
AY358950.1 |
- |
2, 3 |
180 |
|
BC011568.1 |
- |
2, 3 |
202 |
4640182 |
BI561751.1 |
- |
2, 3 |
264 |
|
CB125000.1 |
- |
2, 3 |
234 |
|
KU178743.1 |
- |
1, 2 |
116 |
|
KU178744.1 |
- |
1, 2 |
116 |
|
View Full Product Details