Gene Transcripts

Gene Symbol
SIL1
Entrez Gene ID
64374
Gene Name
SIL1 nucleotide exchange factor
Gene Aliases
BAP, MSS, ULG5
Chromosome Location
Chr. 5: 138946720 - 139198376 on Build GRCh38
UniGene ID
Hs.483521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037633.1 NP_001032722.1 5 517
NM_022464.4 NP_071909.1 4 429
XM_011543570.2 XP_011541872.1 6 554
GenBank mRNA AF547994.1 - 4 348
AJ299442.1 - 4 407
AK074624.1 - 4 421
AK075177.1 - 4 358
AY358950.1 - 4 375
BC011568.1 - 4 397 4640182
BI561751.1 - 4 460
CB125000.1 - 4 429
KU178743.1 - 3 311
KU178744.1 - 3 311

View Full Product Details