Gene Transcripts
- Gene Symbol
- SLC13A3
- Entrez Gene ID
- 64849
- Gene Name
- solute carrier family 13 member 3
- Gene Aliases
- NADC3, SDCT2
- Chromosome Location
- Chr. 20: 46557823 - 46684485 on Build GRCh38
- UniGene ID
- Hs.655498
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001011554.2 |
NP_001011554.1
|
2, 3 |
195 |
|
NM_001193339.1 |
NP_001180268.1
|
1, 2 |
159 |
|
NM_001193340.1 |
NP_001180269.1
|
2, 3 |
195 |
|
NM_022829.5 |
NP_073740.2
|
1, 2 |
159 |
|
GenBank mRNA |
AF154121.1 |
- |
1, 2 |
138 |
|
AK056713.1 |
- |
1, 2 |
157 |
|
AK096658.1 |
- |
2, 3 |
195 |
|
AK295748.1 |
- |
1, 2 |
157 |
|
AK315131.1 |
- |
1, 2 |
157 |
|
AY072810.3 |
- |
1, 2 |
138 |
|
BC014931.1 |
- |
1, 2 |
117 |
|
BC035966.1 |
- |
1, 2 |
157 |
4609776 |
BP358260.1 |
- |
2, 3 |
195 |
|
BQ946167.1 |
- |
2, 3 |
178 |
|
CN310376.1 |
- |
2, 3 |
168 |
|
DA334816.1 |
- |
1, 2 |
157 |
|
DA503319.1 |
- |
2, 3 |
195 |
|
DA636764.1 |
- |
1, 2 |
159 |
|
View Full Product Details