Gene Transcripts

Gene Symbol
HAUS3
Entrez Gene ID
79441
Gene Name
HAUS augmin like complex subunit 3
Gene Aliases
C4orf15, IT1, dgt3
Chromosome Location
Chr. 4: 2231836 - 2242164 on Build GRCh38
UniGene ID
Hs.665869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303143.1 NP_001290072.1 3 1102
NM_024511.6 NP_078787.2 2 831
GenBank mRNA AF040964.1 - 1 575
AK056134.1 - 2 1319
AK293948.1 - 2 1319
BC003648.2 - 3 1103 3534250
BC025356.1 - 1 343 4906369
CR749640.1 - 2 800

View Full Product Details