Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206671.2 | NP_001193600.1 | 2 | 296 | |
NM_001206672.2 | NP_001193601.1 | 2 | 296 | ||
NM_024557.4 | NP_078833.3 | 2 | 296 | ||
NR_045405.1 | 3 | 345 | |||
XM_006718317.3 | XP_006718380.1 | 2 | 341 | ||
XM_006718318.3 | XP_006718381.1 | 2 | 341 | ||
XM_006718321.3 | XP_006718384.1 | 2 | 341 | ||
XM_017018287.1 | XP_016873776.1 | 2 | 341 | ||
XM_017018288.1 | XP_016873777.1 | 3 | 338 | ||
XM_017018289.1 | XP_016873778.1 | 3 | 372 | ||
XM_017018290.1 | XP_016873779.1 | 2 | 341 | ||
GenBank mRNA | AK293933.1 | - | 2 | 259 | |
AK296505.1 | - | 3 | 310 | ||
AK315379.1 | - | 2 | 255 | ||
AM422214.1 | - | 2 | 242 | ||
AY326435.1 | - | 2 | 242 | ||
AY326436.1 | - | 2 | 277 | ||
AY358475.1 | - | 2 | 267 | ||
BC022455.1 | - | 2 | 278 | 4792930 | |
DC420812.1 | - | 2 | 296 | ||
KU178773.1 | - | 2 | 242 | ||
KU178774.1 | - | 2 | 242 |