Gene Transcripts

Gene Symbol
RHBDF2
Entrez Gene ID
79651
Gene Name
rhomboid 5 homolog 2
Gene Aliases
RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2
Chromosome Location
Chr. 17: 76470893 - 76501440 on Build GRCh38
UniGene ID
Hs.464157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005498.3 NP_001005498.2 18 3201
NM_024599.5 NP_078875.4 19 3363
XM_005257669.3 XP_005257726.1 19 3724
XM_005257670.1 XP_005257727.1 19 3233
XM_011525249.2 XP_011523551.1 19 4680
XM_011525250.2 XP_011523552.1 20 3390
XM_011525251.2 XP_011523553.1 19 3207
XM_017025079.1 XP_016880568.1 19 3637
GenBank mRNA AK025994.1 - 14 2387
AK074034.1 - 16 4301
AK225684.1 - 14 2340
AL833680.1 - 12 2421
AY500247.1 - 18 3052
BC016034.1 - 19 3343 3910543
BC035829.1 - 14 2449
BM801831.1 - 1 250

View Full Product Details