Gene Transcripts

Gene Symbol
THAP9
Entrez Gene ID
79725
Gene Name
THAP domain containing 9
Gene Aliases
hTh9
Chromosome Location
Chr. 4: 82900650 - 82920283 on Build GRCh38
UniGene ID
Hs.582050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317776.1 NP_001304705.1 6 1580
NM_024672.5 NP_078948.3 5 1514
XM_005263220.2 XP_005263277.1 5 1471
XM_011532246.2 XP_011530548.1 5 1304
XM_017008603.1 XP_016864092.1 5 2354
XM_017008604.1 XP_016864093.1 5 2407
XM_017008605.1 XP_016864094.1 5 1448
XM_017008606.1 XP_016864095.1 6 1572
XM_017008607.1 XP_016864096.1 5 1787
XM_017008608.1 XP_016864097.1 6 2426
XM_017008609.1 XP_016864098.1 6 1576
XM_017008610.1 XP_016864099.1 6 1563
GenBank mRNA AK026973.1 - 3 844
AK091412.1 - 5 1411
AK225333.1 - 3 851
BC131722.1 - 6 1461
BX648134.1 - 6 1734

View Full Product Details