Gene Transcripts
- Gene Symbol
- FBXL18
- Entrez Gene ID
- 80028
- Gene Name
- F-box and leucine rich repeat protein 18
- Gene Aliases
- Fbl18
- Chromosome Location
- Chr. 7: 5454448 - 5513813 on Build GRCh38
- UniGene ID
- Hs.623974
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057042.1 |
- |
2, 3 |
314 |
|
AK294949.1 |
- |
2, 3 |
354 |
|
DC307306.1 |
- |
2, 3 |
360 |
|
View Full Product Details