Gene Transcripts
- Gene Symbol
- FUZ
- Entrez Gene ID
- 80199
- Gene Name
- fuzzy planar cell polarity protein
- Gene Aliases
- FY, NTD
- Chromosome Location
- Chr. 19: 49806866 - 49817376 on Build GRCh38
- UniGene ID
- Hs.288800
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001171937.1 |
NP_001165408.1
|
3, 4 |
482 |
|
NM_025129.4 |
NP_079405.2
|
4, 5 |
590 |
|
NR_033269.1 |
|
3, 4 |
505 |
|
XM_006723399.3 |
XP_006723462.1
|
4, 5 |
568 |
|
XM_011527339.1 |
XP_011525641.1
|
4, 5 |
568 |
|
XM_011527340.1 |
XP_011525642.1
|
4, 5 |
430 |
|
XM_011527341.2 |
XP_011525643.1
|
4, 5 |
494 |
|
XM_011527342.1 |
XP_011525644.1
|
4, 5 |
717 |
|
XM_011527343.1 |
XP_011525645.1
|
4, 5 |
568 |
|
XM_011527345.1 |
XP_011525647.1
|
4, 5 |
526 |
|
XM_011527346.1 |
XP_011525648.1
|
3, 4 |
444 |
|
XM_011527347.1 |
XP_011525649.1
|
5, 6 |
484 |
|
XM_017027319.1 |
XP_016882808.1
|
4, 5 |
430 |
|
XM_017027320.1 |
XP_016882809.1
|
4, 5 |
568 |
|
XM_017027321.1 |
XP_016882810.1
|
3, 4 |
444 |
|
XM_017027322.1 |
XP_016882811.1
|
4, 5 |
430 |
|
XM_017027323.1 |
XP_016882812.1
|
4, 5 |
568 |
|
GenBank mRNA |
AK026341.1 |
- |
4, 5 |
564 |
|
AK057964.1 |
- |
3, 4 |
385 |
|
AK295072.1 |
- |
4, 5 |
589 |
|
AK315445.1 |
- |
4, 5 |
568 |
|
BC004445.1 |
- |
4, 5 |
491 |
2819805 |
BC010092.2 |
- |
3, 4 |
347 |
3354417 |
BC016793.2 |
- |
4, 5 |
491 |
2819805 |
CB152506.1 |
- |
3, 4 |
468 |
|
View Full Product Details