Gene Transcripts

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
Chromosome Location
Chr. 16: 67109881 - 67148544 on Build GRCh38
UniGene ID
Hs.513666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320540.1 NP_001307469.1 3 217
NM_001320541.1 NP_001307470.1 3 258
NM_001320542.1 NP_001307471.1 3 243
NM_001320543.1 NP_001307472.1 3 258
NM_025187.4 NP_079463.2 2 273
XM_006721284.3 XP_006721347.1 3 241
XM_011523350.2 XP_011521652.1 2 381
XM_017023729.1 XP_016879218.1 3 252
XM_017023730.1 XP_016879219.1 3 255
XM_017023731.1 XP_016879220.1 3 253
GenBank mRNA AF176088.1 - 3 173
AK022138.1 - 1 42
AL540304.3 - 3 152
BC004556.2 - 2 253 3957147
CB997191.1 - 3 216
HY121038.1 - 2 274
HY157930.1 - 3 243

View Full Product Details