Gene Transcripts

Gene Symbol
WDR19
Entrez Gene ID
57728
Gene Name
WD repeat domain 19
Gene Aliases
ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5
Chromosome Location
Chr. 4: 39182404 - 39285810 on Build GRCh38
UniGene ID
Hs.438482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025132.3 NP_079408.3 4, 5 452
XM_011513724.1 XP_011512026.1 4, 5 362
XM_011513725.2 XP_011512027.1 3, 4 317
GenBank mRNA AK026780.1 - 4, 5 327
AK225588.1 - 4, 5 327
AK304161.1 - 4, 5 325
AY029257.1 - 4, 5 452
BC032578.1 - 3, 4 331

View Full Product Details