Gene Transcripts

Gene Symbol
WDR26
Entrez Gene ID
80232
Gene Name
WD repeat domain 26
Gene Aliases
CDW2, GID7, MIP2
Chromosome Location
Chr. 1: 224385143 - 224434299 on Build GRCh38
UniGene ID
Hs.497873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115113.2 NP_001108585.2 14 2791
NM_025160.6 NP_079436.4 14 2839
XM_005273273.4 XP_005273330.1 13 2554
XM_006711817.3 XP_006711880.1 14 2737
XM_011544281.2 XP_011542583.1 14 2580
XM_011544282.2 XP_011542584.1 11 1902
XM_017002396.1 XP_016857885.1 14 2428
GenBank mRNA AB209887.1 - 14 2839
AK024669.1 - 10 1853
AK027236.1 - 1 44
AK225061.1 - 10 1853
AY221751.1 - 14 2629
AY304473.1 - 14 2396
BC031471.2 - 9 1782
BC034498.1 - 16 2831
BC052301.1 - 13 2235 6420398
BC063817.1 - 10 1899
EF011612.1 - 13 2234

View Full Product Details