Gene Transcripts

Gene Symbol
FLAD1
Entrez Gene ID
80308
Gene Name
flavin adenine dinucleotide synthetase 1
Gene Aliases
FAD1, FADS, LSMFLAD, PP591
Chromosome Location
Chr. 1: 154983294 - 154993111 on Build GRCh38
UniGene ID
Hs.118666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184891.1 NP_001171820.1 3 772
NM_001184892.1 NP_001171821.1 3 1411
NM_025207.4 NP_079483.3 2 1216
NM_201398.2 NP_958800.1 3 772
GenBank mRNA AF218022.1 - 3 1338
AF481877.1 - 2 524
AF520568.1 - 2 1137
AK025979.1 - 3 700
AK090542.1 - 2 1158
AK308243.1 - 3 698
BC011378.1 - 3 701 3855223
BC014012.2 - 1 491
BC020253.1 - 1 51
BC021096.1 - 3 690 4891331
BC032323.1 - 3 707 5242727
DQ458779.1 - 2 815
U79241.1 - 1 367

View Full Product Details