Gene Transcripts

Gene Symbol
SLC2A10
Entrez Gene ID
81031
Gene Name
solute carrier family 2 member 10
Gene Aliases
ATS, GLUT10
Chromosome Location
Chr. 20: 46708358 - 46736347 on Build GRCh38
UniGene ID
Hs.305971
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030777.3 NP_110404.1 2 1178
XM_011529060.2 XP_011527362.1 2 1261
XM_011529061.2 XP_011527363.1 2 2818
XM_011529062.2 XP_011527364.1 2 1272
XM_011529063.2 XP_011527365.1 2 1271
XM_011529064.2 XP_011527366.1 2 1273
XM_011529065.2 XP_011527367.1 2 1268
XM_017028087.1 XP_016883576.1 2 1171
GenBank mRNA AF248053.1 - 2 1331
AF321240.1 - 2 1133
AK055548.1 - 2 1178
AK290961.1 - 2 1169
AK315269.1 - 2 1169
AL137188.3 - 1 1089
BC101657.1 - 2 1177 8069163
BC113423.1 - 2 1177 8322475

View Full Product Details