Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030919.2 | NP_112181.3 | 2 | 690 | |
XM_017028088.1 | XP_016883577.1 | 2 | 573 | ||
GenBank mRNA | AK055793.1 | - | 2 | 692 | |
AK303860.1 | - | 3 | 537 | ||
AL832274.1 | - | 2 | 128 | ||
BC001068.2 | - | 2 | 576 | ||
BC006553.2 | - | 2 | 576 | ||
BC053683.1 | - | 2 | 577 | ||
BC063661.1 | - | 2 | 576 | ||
BC080188.1 | - | 2 | 577 |