Gene Transcripts

Gene Symbol
SHCBP1L
Entrez Gene ID
81626
Gene Name
SHC binding and spindle associated 1 like
Gene Aliases
C1orf14, GE36
Chromosome Location
Chr. 1: 182899865 - 182953418 on Build GRCh38
UniGene ID
Hs.497034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030933.2 NP_112195.2 1, 2 689
GenBank mRNA AF288398.1 - 1, 2 688
BC050305.1 - 1, 2 641 4837018
BC132764.1 - 1, 2 665 40146786

View Full Product Details