Gene Transcripts

Gene Symbol
TMPRSS13
Entrez Gene ID
84000
Gene Name
transmembrane protease, serine 13
Gene Aliases
MSP, MSPL, MSPS, TMPRSS11
Chromosome Location
Chr. 11: 117900641 - 117929453 on Build GRCh38
UniGene ID
Hs.266308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077263.2 NP_001070731.1 5 837
NM_001206789.1 NP_001193718.1 4 732
NM_001206790.1 NP_001193719.1 5 837
NM_001244995.1 NP_001231924.1 5 837
GenBank mRNA AB048796.1 - 5 763
AB048797.1 - 4 688
AK027798.1 - 5 781
AK300283.1 - 5 749
AY190317.1 - 5 764
BC018715.1 - 6 572
BC114928.1 - 5 756 40028749

View Full Product Details