Gene Transcripts
- Gene Symbol
- FSD1L
- Entrez Gene ID
- 83856
- Gene Name
- fibronectin type III and SPRY domain containing 1 like
- Gene Aliases
- CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
- Chromosome Location
- Chr. 9: 105442183 - 105552433 on Build GRCh38
- UniGene ID
- Hs.136901
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145313.2 |
NP_001138785.1
|
3, 4 |
402 |
|
NM_001287191.1 |
NP_001274120.1
|
2, 3 |
306 |
|
NM_001287192.1 |
NP_001274121.1
|
2, 3 |
306 |
|
NM_031919.4 |
NP_114125.1
|
2, 3 |
306 |
|
XM_005252254.2 |
XP_005252311.1
|
3, 4 |
635 |
|
XM_005252257.3 |
XP_005252314.1
|
2, 3 |
301 |
|
XM_005252260.3 |
XP_005252317.1
|
2, 3 |
291 |
|
XM_011519077.2 |
XP_011517379.1
|
4, 5 |
293 |
|
XM_011519078.2 |
XP_011517380.2
|
3, 4 |
300 |
|
XM_011519079.2 |
XP_011517381.2
|
3, 4 |
302 |
|
XM_017015182.1 |
XP_016870671.1
|
3, 4 |
635 |
|
XM_017015183.1 |
XP_016870672.1
|
2, 3 |
301 |
|
XM_017015185.1 |
XP_016870674.1
|
3, 4 |
308 |
|
XM_017015187.1 |
XP_016870676.1
|
2, 3 |
301 |
|
GenBank mRNA |
AF088031.1 |
- |
2, 3 |
295 |
|
AF316830.1 |
- |
2, 3 |
169 |
|
AK296433.1 |
- |
2, 3 |
217 |
|
AK299350.1 |
- |
3, 4 |
334 |
|
AK299491.1 |
- |
2, 3 |
217 |
|
AK309817.1 |
- |
3, 4 |
643 |
|
BP219536.1 |
- |
3, 4 |
374 |
|
BP229804.1 |
- |
2, 3 |
306 |
|
BP365251.1 |
- |
2, 3 |
296 |
|
DN999402.1 |
- |
2, 3 |
149 |
|
View Full Product Details