Gene Transcripts

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
Chromosome Location
Chr. 19: 16496311 - 16521369 on Build GRCh38
UniGene ID
Hs.631627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288834.1 NP_001275763.1 2 899
NM_032207.3 NP_115583.1 2 899
XM_006722920.3 XP_006722983.1 2 899
XM_011528354.2 XP_011526656.1 2 899
XM_017027350.1 XP_016882839.1 2 899
GenBank mRNA AK025395.1 - 2 797
AK225118.1 - 2 816
AK225678.1 - 2 809
BC027869.1 - 2 771 5205152

View Full Product Details