Gene Transcripts

Gene Symbol
VPS25
Entrez Gene ID
84313
Gene Name
vacuolar protein sorting 25 homolog
Gene Aliases
DERP9, EAP20, FAP20
Chromosome Location
Chr. 17: 42773436 - 42779600 on Build GRCh38
UniGene ID
Hs.500165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032353.3 NP_115729.1 6 778
GenBank mRNA BC006282.2 - 6 778

View Full Product Details