Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr. 22: 25742133 - 26063847 on Build GRCh38
UniGene ID
Hs.417959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318245.1 NP_001305174.1 38 6219
NM_032608.6 NP_115997.5 38 6216
XM_011530458.2 XP_011528760.1 39 6355
XM_011530459.2 XP_011528761.1 39 6326
XM_011530460.2 XP_011528762.1 38 6199
XM_011530461.2 XP_011528763.1 39 6355
XM_011530464.2 XP_011528766.1 39 6352
XM_011530465.2 XP_011528767.1 38 6112
XM_011530466.2 XP_011528768.1 39 6356
XM_017029012.1 XP_016884501.1 39 6355
XM_017029013.1 XP_016884502.1 39 6403
XM_017029014.1 XP_016884503.1 39 6352
XM_017029015.1 XP_016884504.1 38 6223
XM_017029016.1 XP_016884505.1 27 3479
XM_017029017.1 XP_016884506.1 37 6080
GenBank mRNA AB042648.2 - 34 4528
AB075376.1 - 38 6216
AJ310931.2 - 38 6207
AY077700.1 - 38 6129
BC144596.1 - 36 5871
BC144597.1 - 37 6013 9053127
BC150626.1 - 37 6013 9056996
CR749556.1 - 3 262

View Full Product Details