Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr. 10: 87751512 - 87841345 on Build GRCh38
UniGene ID
Hs.435948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321967.1 NP_001308896.1 9 953
NM_001321968.1 NP_001308897.1 8 863
NM_001321969.1 NP_001308898.1 10 1043
NM_032810.3 NP_116199.2 9 1325
NR_135914.1 10 1040
XM_005270252.4 XP_005270309.1 9 1047
XM_011540302.1 XP_011538604.1 7 718
XM_017016847.1 XP_016872336.1 9 1194
XM_017016848.1 XP_016872337.1 8 1197
XM_017016849.1 XP_016872338.1 10 1166
XM_017016850.1 XP_016872339.1 10 1217
GenBank mRNA AF361493.1 - 8 1159
AK027506.1 - 6 553
AK075223.1 - 9 1254
AK304297.1 - 6 626
AL834370.1 - 8 738
BC010868.1 - 5 388
BC063530.1 - 8 784
BC073998.1 - 9 983 6199813

View Full Product Details