Gene Transcripts

Gene Symbol
KRTAP3-3
Entrez Gene ID
85293
Gene Name
keratin associated protein 3-3
Gene Aliases
KAP3.3, KRTAP3.3
Chromosome Location
Chr. 17: 40993430 - 40994133 on Build GRCh38
UniGene ID
Hs.662759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033185.2 NP_149441.1 1 454
GenBank mRNA AJ406933.1 - 1 454
BC069099.1 - 1 454 7216913

View Full Product Details