Gene Transcripts

Gene Symbol
PURB
Entrez Gene ID
5814
Gene Name
purine rich element binding protein B
Gene Aliases
PURBETA
Chromosome Location
Chr. 7: 44876293 - 44885385 on Build GRCh38
UniGene ID
Hs.349150
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033224.4 NP_150093.1 1 1099
GenBank mRNA AY039216.1 - 1 1074
BU177276.1 - 1 499

View Full Product Details