Gene Transcripts
- Gene Symbol
- CSMD1
- Entrez Gene ID
- 64478
- Gene Name
- CUB and Sushi multiple domains 1
- Gene Aliases
- PPP1R24
- Chromosome Location
- Chr. 8: 2935353 - 4995035 on Build GRCh38
- UniGene ID
- Hs.571466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033225.5 |
NP_150094.5
|
22, 23 |
3872 |
|
XM_011534752.2 |
XP_011533054.1
|
22, 23 |
4101 |
|
XM_011534753.2 |
XP_011533055.1
|
5, 6 |
979 |
|
XM_017013731.1 |
XP_016869220.1
|
22, 23 |
4101 |
|
GenBank mRNA |
AB067477.1 |
- |
3, 4 |
590 |
|
AB209502.1 |
- |
10, 11 |
1924 |
|
AF333704.2 |
- |
23, 24 |
3875 |
|
AK126857.1 |
- |
13, 14 |
2044 |
|
AY017307.1 |
- |
23, 24 |
3770 |
|
AY358174.1 |
- |
6, 7 |
1015 |
|
View Full Product Details