Gene Transcripts

Gene Symbol
RBP7
Entrez Gene ID
116362
Gene Name
retinol binding protein 7
Gene Aliases
CRBP4, CRBPIV
Chromosome Location
Chr. 1: 9997197 - 10016020 on Build GRCh38
UniGene ID
Hs.422688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052960.2 NP_443192.1 3 348
GenBank mRNA AF399927.1 - 3 329
AK312027.1 - 3 316
AY145438.1 - 3 315
BC033883.1 - 4 561
BC063013.1 - 3 348 5224517

View Full Product Details