Gene Transcripts

Gene Symbol
CST9L
Entrez Gene ID
128821
Gene Name
cystatin 9-like
Gene Aliases
CTES7B, bA218C14.1
Chromosome Location
Chr. 20: 23564732 - 23568749 on Build GRCh38
UniGene ID
Hs.121554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080610.2 NP_542177.1 1 496
GenBank mRNA AK312112.1 - 1 494
AY358615.1 - 1 439
BC029656.1 - 1 496 5163974

View Full Product Details