Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080867.2 | NP_543143.1 | 2 | 632 | |
NM_199421.1 | NP_955453.1 | 3 | 761 | ||
XM_011536425.1 | XP_011534727.1 | 3 | 815 | ||
XM_011536426.1 | XP_011534728.1 | 3 | 730 | ||
GenBank mRNA | AF424815.1 | - | 1 | 412 | |
AK124083.1 | - | 2 | 485 | ||
BC060790.1 | - | 3 | 639 | 30338646 | |
BX161418.1 | - | 2 | 528 | ||
BX648734.1 | - | 2 | 418 | ||
CD109883.1 | - | 2 | 659 | ||
CR933659.1 | - | 2 | 522 |