Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005859.4 | NP_005850.1 | 1 | 895 | |
GenBank mRNA | BC036087.1 | - | 1 | 848 | |
BC110574.1 | - | 2 | 725 | ||
BC110594.1 | - | 2 | 774 | ||
BT019388.1 | - | 1 | 836 | ||
M96684.1 | - | 1 | 895 |