Gene Transcripts

Gene Symbol
PRELID2
Entrez Gene ID
153768
Gene Name
PRELI domain containing 2
Gene Aliases
Chromosome Location
Chr. 5: 145587325 - 145835369 on Build GRCh38
UniGene ID
Hs.314261
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138492.5 NP_612501.3 6, 7 803
NM_182960.3 NP_892005.1 6, 7 636
NM_205846.2 NP_995318.1 5, 6 600
XM_017009129.1 XP_016864618.1 6, 7 636
XM_017009130.1 XP_016864619.1 6, 7 636
XM_017009131.1 XP_016864620.1 6, 7 636
GenBank mRNA AK095695.1 - 6, 7 571
BC012033.1 - 6, 7 770 4476359
BC028175.1 - 517
BQ645893.1 - 5, 6 570
BU167299.1 - 5, 6 462
BU186346.1 - 6, 7 573

View Full Product Details