Gene Transcripts

Gene Symbol
STXBP5
Entrez Gene ID
134957
Gene Name
syntaxin binding protein 5
Gene Aliases
LGL3, LLGL3, Nbla04300
Chromosome Location
Chr. 6: 147204184 - 147390476 on Build GRCh38
UniGene ID
Hs.736439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127715.2 NP_001121187.1 24 2741
NM_139244.4 NP_640337.3 22 2633
XM_006715339.2 XP_006715402.1 23 2867
XM_011535453.1 XP_011533755.1 23 2465
XM_017010267.1 XP_016865756.1 19 2812
GenBank mRNA AB073398.1 - 13 1472
AK055484.1 - 5 451
AK090549.1 - 14 1451
AK128121.1 - 23 2460
AL834152.1 - 25 2790
BC113382.1 - 22 2506 8322434
BC113408.1 - 22 2506 8322460

View Full Product Details