Gene Transcripts

Gene Symbol
MIEF2
Entrez Gene ID
125170
Gene Name
mitochondrial elongation factor 2
Gene Aliases
MID49, SMCR7
Chromosome Location
Chr. 17: 18260534 - 18265790 on Build GRCh38
UniGene ID
Hs.655555
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144900.1 NP_001138372.1 3 426
NM_139162.3 NP_631901.2 3 426
NM_148886.1 NP_683684.2 3 274
XM_017024190.1 XP_016879679.1 3 771
GenBank mRNA AF467443.1 - 3 274
AK056165.1 - 3 405
AK128310.1 - 3 329
BC014973.1 - 3 274 4859126
DB183618.1 - 3 432

View Full Product Details