Gene Transcripts

Gene Symbol
TMEM42
Entrez Gene ID
131616
Gene Name
transmembrane protein 42
Gene Aliases
Chromosome Location
Chr. 3: 44861909 - 44865667 on Build GRCh38
UniGene ID
Hs.646856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144638.2 NP_653239.1 3 911
GenBank mRNA AL532443.3 - 4 903
AL834253.1 - 2 2996
BC019851.1 - 3 904 5114515
BX103899.1 - 1 496

View Full Product Details