Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr. 12: 8822472 - 8887202 on Build GRCh38
UniGene ID
Hs.620532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282424.2 NP_001269353.1 23 3005
NM_144670.5 NP_653271.2 34 4490
XM_011520566.2 XP_011518868.1 34 4447
XM_011520567.2 XP_011518869.1 34 4447
XM_017018868.1 XP_016874357.1 34 4444
XM_017018869.1 XP_016874358.1 34 4444
XM_017018870.1 XP_016874359.1 34 4408
GenBank mRNA AK057908.1 - 5 492
AK096448.1 - 12 1479
AK122624.1 - 28 3576
AK123591.1 - 17 2143
AK123592.1 - 23 2960
AK123601.1 - 31 3867
AK290355.1 - 34 4343
AK302555.1 - 23 3005
AL832139.1 - 34 4341
AL832750.1 - 12 1215
AL833479.1 - 32 4006
BC093840.1 - 5 443
BC112131.1 - 5 443
BX647674.1 - 16 1945
BX648231.1 - 35 4342

View Full Product Details