Gene Transcripts

Gene Symbol
ARHGAP36
Entrez Gene ID
158763
Gene Name
Rho GTPase activating protein 36
Gene Aliases
Chromosome Location
Chr. X: 131058242 - 131089885 on Build GRCh38
UniGene ID
Hs.22905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282607.1 NP_001269536.1 4, 5 673
NM_144967.3 NP_659404.2 4, 5 909
XM_006724729.1 XP_006724792.1 3, 4 380
XM_011531280.1 XP_011529582.1 3, 4 397
GenBank mRNA AK054620.1 - 1, 2 269
AK128203.1 - 4, 5 909
AK294274.1 - 4, 5 632
AK296748.1 - 4, 5 651
BC063790.1 - 4, 5 508 6196091

View Full Product Details