Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304492.1 | NP_001291421.1 | 4, 5 | 388 | |
NM_145911.2 | NP_666016.1 | 5, 6 | 960 | ||
GenBank mRNA | AF507946.1 | - | 5, 6 | 951 | |
AK091039.1 | - | 4, 5 | 383 | ||
BC007256.1 | - | 5, 6 | 951 | 3028040 | |
BT007400.1 | - | 2, 3 | 137 |