Gene Transcripts

Gene Symbol
TTC30A
Entrez Gene ID
92104
Gene Name
tetratricopeptide repeat domain 30A
Gene Aliases
IFT70A
Chromosome Location
Chr. 2: 177614298 - 177618966 on Build GRCh38
UniGene ID
Hs.128384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152275.3 NP_689488.3 1 2432
GenBank mRNA AK057530.1 - 2 2069
AK125104.1 - 2 2136
AK292395.1 - 1 2416
AL049329.1 - 1 532
BC042848.1 - 1 2436 4831326
BC048006.1 - 1 2206 5299020

View Full Product Details