Gene Transcripts

Gene Symbol
LRR1
Entrez Gene ID
122769
Gene Name
leucine rich repeat protein 1
Gene Aliases
4-1BBLRR, LRR-1, PPIL5
Chromosome Location
Chr. 14: 49598697 - 49614672 on Build GRCh38
UniGene ID
Hs.451090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152329.3 NP_689542.2 2 584
NM_203467.1 NP_982292.1 2 584
NR_037792.1 2 584
NR_037793.1 3 692
GenBank mRNA AK293156.1 - 2 303
AY052405.1 - 2 270
BC030142.2 - 2 520
BC064133.1 - 3 437
BC093697.1 - 2 286
BC107888.1 - 2 333
BC112241.1 - 2 286
BC139921.1 - 2 324 40148567
BI825814.1 - 2 511
BI826606.1 - 2 585
BI829295.1 - 2 521
BQ059807.1 - 2 254
BX248298.1 - 2 327
BX648029.1 - 2 507

View Full Product Details