Gene Transcripts

Gene Symbol
SPATA2L
Entrez Gene ID
124044
Gene Name
spermatogenesis associated 2 like
Gene Aliases
C16orf76, tamo
Chromosome Location
Chr. 16: 89696357 - 89701993 on Build GRCh38
UniGene ID
Hs.374556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152339.3 NP_689552.2 3 1944
XM_005256279.4 XP_005256336.1 3 2361
GenBank mRNA AF070574.1 - 1 1175
AK127372.1 - 2 1748
BC027606.1 - 4 611
BC039608.1 - 3 1922 5248678
BC053588.1 - 3 1926 5245683

View Full Product Details