Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000362.4 | NP_000353.1 | 3 | 1482 | |
GenBank mRNA | AK298126.1 | - | 4 | 302 | |
AK314871.1 | - | 3 | 591 | ||
BC014277.2 | - | 3 | 539 | 3689386 | |
BT006848.1 | - | 3 | 296 | ||
CR456593.1 | - | 3 | 344 | ||
L15078.1 | - | 3 | 254 | ||
S78453.1 | - | 3 | 572 | ||
U02571.1 | - | 3 | 577 | ||
U14394.1 | - | 3 | 583 | ||
U67195.1 | - | 3 | 574 | ||
X76227.1 | - | 3 | 366 |