Gene Transcripts

Gene Symbol
DNAJB8
Entrez Gene ID
165721
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
Gene Aliases
CT156, DJ6
Chromosome Location
Chr. 3: 128462432 - 128467248 on Build GRCh38
UniGene ID
Hs.518241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153330.4 NP_699161.1 3 1822
XM_006713519.3 XP_006713582.1 2 2719
GenBank mRNA AK125946.1 - 2 2719
AK126068.1 - 1 397
BC029521.1 - 1 415 5296554
BC050288.1 - 3 1822 5270636

View Full Product Details