Gene Transcripts

Gene Symbol
TAPT1
Entrez Gene ID
202018
Gene Name
transmembrane anterior posterior transformation 1
Gene Aliases
CMVFR, OCLSBG
Chromosome Location
Chr. 4: 16160505 - 16226538 on Build GRCh38
UniGene ID
Hs.479223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153365.2 NP_699196.2 2 304
XM_005248139.2 XP_005248196.1 2 237
XM_005248140.3 XP_005248197.1 2 187
XM_011513812.2 XP_011512114.1 2 302
XM_011513815.2 XP_011512117.1 2 1522
XM_017007876.1 XP_016863365.1 2 128
GenBank mRNA AK074494.1 - 2 237
BC066899.1 - 2 242 4825556
DB461077.1 - 2 307

View Full Product Details