Gene Transcripts
- Gene Symbol
- ALDH5A1
- Entrez Gene ID
- 7915
- Gene Name
- aldehyde dehydrogenase 5 family member A1
- Gene Aliases
- SSADH, SSDH
- Chromosome Location
- Chr. 6: 24494969 - 24537207 on Build GRCh38
- UniGene ID
- Hs.371723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080.3 |
NP_001071.1
|
1, 2 |
390 |
|
NM_170740.1 |
NP_733936.1
|
1, 2 |
390 |
|
GenBank mRNA |
AJ427353.1 |
- |
1, 2 |
158 |
|
AJ427354.2 |
- |
1, 2 |
158 |
|
AJ427355.2 |
- |
1, 2 |
362 |
|
AK290800.1 |
- |
1, 2 |
508 |
|
AK294699.1 |
- |
2, 3 |
167 |
|
AK314357.1 |
- |
1, 2 |
390 |
|
AK315380.1 |
- |
1, 2 |
485 |
|
BC034321.1 |
- |
1, 2 |
390 |
4829533 |
KJ534766.1 |
- |
1, 2 |
362 |
|
KJ534767.1 |
- |
1, 2 |
362 |
|
KJ534768.1 |
- |
1, 2 |
362 |
|
KJ534769.1 |
- |
1, 2 |
362 |
|
Y11192.1 |
- |
1, 2 |
362 |
|
View Full Product Details