Gene Transcripts
- Gene Symbol
- SKP1
- Entrez Gene ID
- 6500
- Gene Name
- S-phase kinase-associated protein 1
- Gene Aliases
- EMC19, OCP-II, OCP2, SKP1A, TCEB1L, p19A
- Chromosome Location
- Chr. 5: 134156391 - 134177033 on Build GRCh38
- UniGene ID
- Hs.171626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006930.3 |
NP_008861.2
|
3, 4 |
358 |
|
NM_170679.2 |
NP_733779.1
|
3, 4 |
358 |
|
GenBank mRNA |
BC009839.2 |
- |
3, 4 |
242 |
|
BC020798.1 |
- |
3, 4 |
293 |
4243711 |
BC025673.1 |
- |
3, 4 |
255 |
5203008 |
BC065730.1 |
- |
3, 4 |
279 |
6672613 |
BI552966.1 |
- |
3, 4 |
325 |
|
BI670085.1 |
- |
3, 4 |
363 |
|
U33760.1 |
- |
3, 4 |
272 |
|
U37558.1 |
- |
2, 3 |
140 |
|
Z47087.1 |
- |
3, 4 |
288 |
|
- Gene Symbol
- SKP1P2
- Entrez Gene ID
- 728622
- Gene Name
- S-phase kinase-associated protein 1 pseudogene 2
- Gene Aliases
- SKP1B, p19B
- Chromosome Location
- Chr. 12: 16988747 - 16990628 on Build GRCh38
- UniGene ID
- Hs.741380
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036619.1 |
|
1 |
558 |
|
GenBank mRNA |
AK310444.1 |
- |
1 |
558 |
|
View Full Product Details