Gene Transcripts

Gene Symbol
CSN1S2AP
Entrez Gene ID
286828
Gene Name
casein alpha s2-like A, pseudogene
Gene Aliases
CSN1S2A
Chromosome Location
Chr. 4: 70067386 - 70085273 on Build GRCh38
UniGene ID
Hs.631945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003720.1 8 265
GenBank mRNA AY154892.1 - 8 265
BC128170.1 - 5 139
BC128173.1 - 5 139
BC141914.1 - 5 139
BC150502.1 - 5 139

View Full Product Details